Learn More
4-Chlorobenzotrichloride, TRC
Product Code. 30363889
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363889 5 g -
30348799 25 g -
30365715 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363889 Supplier Toronto Research Chemicals Supplier No. TRCC3475855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5216-25-1
Molecular Formula C7 H4 Cl4
Molecular Weight (g/mol) 229.92
Synonym alpha,alpha,alpha,4-Tetrachlorotoluene, Benzene, 1-chloro-4-(trichloromethyl)-, Toluene, p,α,α,α-tetrachloro- (6CI,7CI,8CI), 1-Chloro-4-(trichloromethyl)benzene, 4,α,α,α-Tetrachlorotoluene, 4-Chlorobenzotrichloride, p,α,α,α-Tetrachlorotoluene, p-(Trichloromethyl)chlorobenzene, p-Chloro-α,α,α-trichlorotoluene, p-Chlorobenzotrichloride, p-Chlorophenyltrichloromethane, α,α,α,4-Tetrachlorotoluene, α,α,α,p-Tetrachlorotoluene, α,α,α-Trichloro-4-chlorotoluene
IUPAC Name 1-chloro-4-(trichloromethyl)benzene
SMILES Clc1ccc(cc1)C(Cl)(Cl)Cl
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H4Cl4/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H
Formula Weight 227.9067
Chemical Name or Material alpha,alpha,alpha,4-Tetrachlorotoluene