missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzophenone, TRC

Product Code. 30012920
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012920 1 g 1g
30133817 10 g 10g
30003701 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012920 Supplier Toronto Research Chemicals Supplier No. TRCC3644851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-85-0
Formula Weight 216.0342
InChI Formula InChI=1S/C13H9ClO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H
IUPAC Name (4-chlorophenyl)-phenylmethanone
Molecular Formula C13 H9 Cl O
Molecular Weight (g/mol) 216.66
Percent Purity >95%
SMILES Clc1ccc(cc1)C(=O)c2ccccc2
Synonym (4-Chlorophenyl)phenylmethanone, 4-Chlorobenzophenone, Meclozine Hydrochloride Imp. C (EP)
Chemical Name or Material 4-Chlorobenzophenone
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.