missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylbenzenesulphonamide, TRC

Product Code. 30367529
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367529 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367529 Supplier Toronto Research Chemicals Supplier No. TRCC42848050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-N-methylbenzenesulphonamide
CAS 6333-79-5
Molecular Formula C7H8ClNO2S
Quantity 50 mg
Synonym 1-[(N-Methylamino)sulfonyl]-4-chloro-benzene, N-Methyl-4-chlorobenzenesulfonamide, N-Methyl-p-chlorobenzenesulfonamide, NSC 31051, NSC 404320
InChI Formula InChI=1S/C7H8ClNO2S/c1-9-12(10,11)7-4-2-6(8)3-5-7/h2-5,9H,1H3
SMILES CNS(=O)(=O)c1ccc(Cl)cc1
IUPAC Name 4-chloro-N-methylbenzenesulfonamide
Molecular Weight (g/mol) 205.66
Formula Weight 204.9964
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.