missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoyl Chloride (>90%), TRC

Product Code. 30351741
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351741 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351741 Supplier Toronto Research Chemicals Supplier No. TRCC4281405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-3-sulfamoylbenzoyl Chloride
CAS 70049-77-3
Molecular Formula C7H5Cl2NO3S
Quantity 5 g
Synonym 3-(Aminosulfonyl)-4-chlorobenzoyl Chloride
InChI Formula InChI=1S/C7H5Cl2NO3S/c8-5-2-1-4(7(9)11)3-6(5)14(10,12)13/h1-3H,(H2,10,12,13)
SMILES NS(=O)(=O)c1cc(ccc1Cl)C(=O)Cl
IUPAC Name 4-chloro-3-sulfamoylbenzoyl chloride
Molecular Weight (g/mol) 254.09
Formula Weight 252.9367
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.