Learn More
4-Bromobenzenesulfonyl chloride, 98+%
Description
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.
Solubility
Soluble in Tetrahydrofuran, petroleum ether and 1,4-dioxane. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas.
Specifications
Specifications
| Melting Point | 73°C to 77°C |
| Boiling Point | 152°C to 154°C (26 mmHg) |
| Quantity | 100 g |
| UN Number | UN3261 |
| Beilstein | 743518 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,1407 |
| Solubility Information | Soluble in Tetrahydrofuran,petroleum ether and 1,4-dioxane. Insoluble in water. |
| Formula Weight | 255.52 |
| Percent Purity | ≥98% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.