missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-chloroacetanilide, 98%, Thermo Scientific™

Product Code. 11477867
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11477867

Brand: Thermo Scientific Alfa Aesar A16547.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 22459-81-0
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040848
InChI Key BVADKOYGDKTWMU-UHFFFAOYSA-N
Synonym 4'-bromo-3'-chloroacetanilide, n-4-bromo-3-chlorophenyl acetamide, n-acetyl 4-bromo-3-chloroaniline, 4-bromo-3-chloroacetanilide, n1-4-bromo-3-chlorophenyl acetamide, acetamide,n-4-bromo-3-chlorophenyl, acmc-209fwl, maybridge1_008946, n-acetyl4-bromo-3-chloroaniline
PubChem CID 2746295
IUPAC Name N-(4-bromo-3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)Cl

Tekniske data

Melting Point 123°C to 125°C
Quantity 25 g
Beilstein 2967800
Formula Weight 248.51
Percent Purity 98%
Chemical Name or Material 4'-Bromo-3'-chloroacetanilide
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.