Learn More
4-(Boc-amino)piperidine, 96%
Product Code. 11452070
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452070 25 g 25g
11432070 1 g 1g
11442070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11452070 Supplier Thermo Scientific Alfa Aesar Supplier No. H28517.14
Request Bulk or Custom Format
CAS: 73874-95-0 | C10H21N2O2 | 201.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73874-95-0
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD00798171
InChI Key CKXZPVPIDOJLLM-UHFFFAOYSA-O
Synonym 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
PubChem CID 723833
SMILES CC(C)(C)OC(=O)NC1CC[NH2+]CC1
Specifications
Melting Point 162°C to 166°C
IUPAC Name 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium
Quantity 25 g
Formula Weight 200.28
Percent Purity 96%
Chemical Name or Material 4-(Boc-amino)piperidine