missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-L-phenylalanine, 95%

Product Code. p-7034399
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408617 250 mg 250mg
15488607 1 g 1g
15498607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408617 Supplier Thermo Scientific Alfa Aesar Supplier No. H52083.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104504-45-2 | C16H15NO3 | 269.30 g/mol

4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104504-45-2
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00238102
InChI Key TVIDEEHSOPHZBR-AWEZNQCLSA-N
Synonym s-2-amino-3-4-benzoylphenyl propanoic acid, l-4-benzoylphenylalanine, para-benzoyl-phenylalanine, h-bpa-oh h-phe 4-bz-oh, 4-benzoyl-l-phenylalanine, 4-benzoylphenylalanine, l-p-benzoylphenylalanine, phenylalanine, 4-benzoyl, h-bpa-oh, h-p-bz-phe-oh
PubChem CID 7020128
IUPAC Name (2S)-2-amino-3-(4-benzoylphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 269.3
Percent Purity 95%
Chemical Name or Material 4-Benzoyl-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.