Learn More
4-Aminophenylphosphorylcholine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminophenylphosphorylcholine |
| CAS | 102185-28-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11 H19 N2 O4 P |
| Quantity | 250 mg |
| Synonym | Ethanaminium, 2-[[(4-aminophenoxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt, 2-[[(4-Aminophenoxy)hydroxyphosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, 4-Aminophenylphosphorylcholine |
| InChI Formula | InChI=1S/C11H19N2O4P/c1-13(2,3)8-9-16-18(14,15)17-11-6-4-10(12)5-7-11/h4-7H,8-9,12H2,1-3H3 |
| SMILES | C[N+](C)(C)CCOP(=O)([O-])Oc1ccc(N)cc1 |
| IUPAC Name | (4-aminophenyl) 2-(trimethylazaniumyl)ethyl phosphate |
| Molecular Weight (g/mol) | 274.25 |
| Show More |