missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenyl 2-Acetamido-2-deoxy-Alpha-D-galactopyranoside Hydrochloride, TRC

Product Code. 30326723
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326723 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326723 Supplier Toronto Research Chemicals Supplier No. TRCA6220005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminophenyl 2-Acetamido-2-deoxy-a-D-galactopyranoside, Hyd
CAS 210049-16-4
Molecular Formula C14 H20 N2 O6 . Cl H
Quantity 5 mg
InChI Formula InChI=1S/C14H20N2O6.ClH/c1-7(18)16-11-13(20)12(19)10(6-17)22-14(11)21-9-4-2-8(15)3-5-9;/h2-5,10-14,17,19-20H,6,15H2,1H3,(H,16,18);1H/t10-,11-,12+,13-,14+;/m1./s1
SMILES Cl.CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccc(N)cc2
IUPAC Name N-[(2R,3R,4R,5R,6R)-2-(4-aminophenoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide;hydrochloride
Molecular Weight (g/mol) 348.78
Formula Weight 348.1088
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.