missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%

Product Code. p-4228306 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10775052 5 g Glass bottle 5g
10391800 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10775052 Supplier Thermo Scientific Acros Supplier No. 251850050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-30-2
Molecular Formula C6H8ClN3O4S2
Molecular Weight (g/mol) 285.73
InChI Key IHJCXVZDYSXXFT-UHFFFAOYSA-N
Synonym 4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
PubChem CID 67136
ChEBI CHEBI:3602
IUPAC Name 4-amino-6-chlorobenzene-1,3-disulfonamide
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 260.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,2076
Formula Weight 285.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Amino-6-chloro-1, 3-benzenedisulfonamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.