missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4' '-Amino-5-O-[(2-propenyloxy)carbonyl] O’-TMS Abamectin, TRC
Specifications
Specifications
| Analyte or Component Names | 4' '-Amino-5-O-((2-propenyloxy)carbonyl) O'-TMS Abamectin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C55 H85 N O15 Si |
| InChI Formula | InChI=1S/C55H85NO15Si/c1-15-24-61-53(58)68-49-35(7)25-41-52(57)65-40-26-39(69-54(29-40)23-22-34(6)48(70-54)31(3)16-2)21-20-33(5)47(32(4)18-17-19-38-30-62-51(49)55(38,41)71-72(12,13)14)66-45-28-43(60-11)50(37(9)64-45)67-44-27-42(59-10)46(56)36(8)63-44/h15,17-20,22-23,25,31-32,34,36-37,39-51H,1,16,21,24,26-30,56H2,2-14H3/b18-17+,33-20+,38-19-/t31-,32-,34-,36-,37-,39+,40-,41-,42-,43-,44-,45-,46+,47-,48+,49+,50-,51+,54+,55+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@H](N)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C/6\CO[C@@H]7[C@H](OC(=O)OCC=C)C(=C[C@@H](C(=O)O3)[C@]67O[Si](C)(C)C)C)O2)C=C[C@@H]1C |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 1028.35 |
| Quantity | 2.5 mg |
| Formula Weight | 1027.5688 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?