missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-chloro-2-methoxybenzoic Acid, TRC

Product Code. 17961785
Change view
Click to view available options
Quantity:
250 mg
1 g
50 g
Unit Size:
1g
250mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961785 1 g 1g
30132443 50 g 50g
17931655 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961785 Supplier Toronto Research Chemicals Supplier No. TRCA6033201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7206-70-4
Formula Weight 201.0193
InChI Formula InChI=1S/C8H8ClNO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,10H2,1H3,(H,11,12)
IUPAC Name 4-amino-5-chloro-2-methoxybenzoic acid
Molecular Formula C8 H8 Cl N O3
Molecular Weight (g/mol) 201.61
SMILES COc1cc(N)c(Cl)cc1C(=O)O
Synonym 4-Amino-5-chloro-2-methoxybenzoic Acid, Metoclopramide Imp. C (EP)
Chemical Name or Material 4-Amino-5-chloro-2-methoxybenzoic Acid

Specifications

Analyte or Component Names 4-Amino-5-chloro-2-methoxybenzoic Acid
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.