missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxybutyric acid

Product Code. p-7043029
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15486749

Brand: Thermo Scientific Alfa Aesar J65145.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 924-49-2 | C4H9NO3 | 119.12 g/mol

Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reduces uptake of iodine by the thyroid glands in animals
  • Antispasmodic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-49-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00008141
InChI Key YQGDEPYYFWUPGO-UHFFFAOYSA-N
Synonym 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal
PubChem CID 2149
ChEBI CHEBI:16080
IUPAC Name 4-amino-3-hydroxybutanoic acid
SMILES C(C(CN)O)C(=O)O

Specifications

Melting Point 223°C (decomposition)
Color White to Yellow
Beilstein 1721708
Merck Index 14,444
Quantity 5 g
Formula Weight 119.12
Physical Form Powder
Chemical Name or Material 4-Amino-3-hydroxybutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.