missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-4-methylpimelic acid, 97%, Thermo Scientific™

Product Code. 11431357
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11431357 1 g 1g
11441357 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11431357

Brand: Thermo Scientific Alfa Aesar B24891.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19830-09-2
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.22
MDL Number MFCD00052871
InChI Key LYBQMKJABQTTFJ-UHFFFAOYSA-L
Synonym 4-acetyl-4-methylpimelic acid, aav2ytm2mi, unii-aav2ytm2mi, 3-acetyl-3-methylpentane-1,5-dicarboxylic acid, acmc-1c6z0, 4-acetyl-4-methyl-heptanedioic acid, heptanedioic acid,4-acetyl-4-methyl
PubChem CID 407651
IUPAC Name 4-acetyl-4-methylheptanedioic acid
SMILES CC(=O)C(C)(CCC([O-])=O)CCC([O-])=O

Specifications

Melting Point 121°C to 125°C
Quantity 1 g
Beilstein 1789115
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 4-Acetyl-4-methylpimelic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.