Learn More
4-Acetamidophenyl Beta-D-Glucuronide Sodium Salt, TRC
Product Code. 30135266
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135266 50 mg 50mg
30009462 10 mg 10mg
30008870 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135266 Supplier Toronto Research Chemicals Supplier No. TRCA15850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120595-80-4
Molecular Formula C14 H16 N O8 . Na
Molecular Weight (g/mol) 349.27
Synonym β-D-Glucopyranosiduronic acid, 4-(acetylamino)phenyl, sodium salt (1:1), β-D-Glucopyranosiduronic acid, 4-(acetylamino)phenyl, monosodium salt (9CI), 4-Acetamidophenyl β-D-glucuronide sodium salt, N-Acetyl-4-aminophenol glucuronide sodium salt, Acetaminophen glucuronide sodium salt, Paracetamol glucuronide sodium, Paracetamol β-D-glucuronide sodium salt, Paracetamol O-glucuronide sodium
IUPAC Name sodium;(2S,3S,4S,5R,6S)-6-(4-acetamidophenoxy)-3,4,5-trihydroxyoxane-2-carboxylate
SMILES [Na+].CC(=O)Nc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-])cc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H17NO8.Na/c1-6(16)15-7-2-4-8(5-3-7)22-14-11(19)9(17)10(18)12(23-14)13(20)21;/h2-5,9-12,14,17-19H,1H3,(H,15,16)(H,20,21);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1
Formula Weight 349.0774
Percent Purity >95%
Chemical Name or Material 4-Acetamidophenyl beta-D-Glucuronide, Sodium Salt
Grade HPLC