missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenyl Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30009462
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009462 10 mg 10mg
30135266 50 mg 50mg
30008870 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009462 Supplier Toronto Research Chemicals Supplier No. TRCA15850010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120595-80-4
Molecular Formula C14 H16 N O8 . Na
Molecular Weight (g/mol) 349.27
Synonym β-D-Glucopyranosiduronic acid, 4-(acetylamino)phenyl, sodium salt (1:1), β-D-Glucopyranosiduronic acid, 4-(acetylamino)phenyl, monosodium salt (9CI), 4-Acetamidophenyl β-D-glucuronide sodium salt, N-Acetyl-4-aminophenol glucuronide sodium salt, Acetaminophen glucuronide sodium salt, Paracetamol glucuronide sodium, Paracetamol β-D-glucuronide sodium salt, Paracetamol O-glucuronide sodium
IUPAC Name sodium;(2S,3S,4S,5R,6S)-6-(4-acetamidophenoxy)-3,4,5-trihydroxyoxane-2-carboxylate
SMILES [Na+].CC(=O)Nc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-])cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H17NO8.Na/c1-6(16)15-7-2-4-8(5-3-7)22-14-11(19)9(17)10(18)12(23-14)13(20)21;/h2-5,9-12,14,17-19H,1H3,(H,15,16)(H,20,21);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1
Formula Weight 349.0774
Percent Purity >95%
Chemical Name or Material 4-Acetamidophenyl beta-D-Glucuronide, Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.