missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-6 wt% Soot in Diesel Engine Oil, VHG™
Description
4-6 wt% Soot in Diesel Engine Oil
Specifications
Specifications
| Chemical Name or Material | Nominal Soot Content Range |
| Methods | FTIR/IR |
| Analyte or Component Names | Nominal Soot Content Range |
| Shipping Condition | Room Temperature |
| IUPAC Name | Nominal Soot Content Range |
| Solvent or Matrix | 15W40 Motor Oil |
| Quantity | 50 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?