missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dioxoheptanoic acid, MedChemExpress

Product Code. 16455447
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16455447

Brand: Medchem Express HYW010184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4,6-Dioxoheptanoic acid is a potent inhibitor of heme biosynthesis.

4,6-Dioxoheptanoic acid is a potent inhibitor of heme biosynthesis.

Specifications

Chemical Name or Material 4,6-Dioxoheptanoic acid
CAS 51568-18-4
Color White
Purity Grade Notes Research
Molecular Formula C7H10O4
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (632.31 mM; Need ultrasonic)
SMILES CC(=O)CC(=O)CCC(O)=O
Molecular Weight (g/mol) 158.15
Formula Weight 158.15
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.