missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydroxy-1,3-benznedisulphonic acid disodium salt, 97.4%, MP Biomedicals™

Product Code. 11491141 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11491141

Brand: MP Biomedicals 0215774125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym tiron, tiferron, sodium catechol sulfate, disodium 4,5-dihydroxybenzene-1,3-disulfonate, 3,5-disulfocatechol disodium salt, sodium pyrocatechol-3,5-disulfonate, disodium pyrocatechol-3,5-disulfonate, sodium 1,2-dihydroxybenzenedisulfonate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid, 4,5-dihydroxy-, disodium salt
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Color Tan to White
Assay Percent Range 97.4%
Quantity 25 g
Formula Weight 332.22
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Dihydroxy-1, 3-benznedisulphonic acid disodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.