missing translation for 'onlineSavingsMsg'
Learn More

4,5,6-Triaminopyrimidine sulfate hydrate, 98+% (dry wt.), water <8%

Product Code. p-7025724
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11409317

Brand: Thermo Scientific Alfa Aesar B21369.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207742-76-5 | C4H9N5O4S | 223.21 g/mol

4,5,6-Triaminopyrimidine sulfate hydrate is used to prepare 2,5-anhydro-3-deoxy-N-(4',6'-diaminopyrimidin-5'yl)-D-ribo-hexonamide by reaction with 2,5-anhydro-4-O-benzyl-3-deoxy-D-ribo-hexonic acid. It is also used to prepare 4,5,6-triaminopyrimidine sulfite. Further, it acts as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5,6-Triaminopyrimidine sulfate hydrate is used to prepare 2,5-anhydro-3-deoxy-N-(4′,6′-diaminopyrimidin-5′yl)-D-ribo-hexonamide by reaction with 2,5-anhydro-4-O-benzyl-3-deoxy-D-ribo-hexonic acid. It is also used to prepare 4,5,6-triaminopyrimidine sulfite. Further, it acts as an intermediate in synthetic chemistry.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207742-76-5
Molecular Formula C4H9N5O4S
Molecular Weight (g/mol) 223.21
MDL Number MFCD00012789
InChI Key RKJICTKHLYLPLY-UHFFFAOYSA-N
Synonym 4,5,6-triaminopyrimidine sulfate hydrate, 4,5,6-triaminepyrimidine sulfate hydrate, pyrimidine, 4,5,6-triamino-, sulfate, hydrate, pyrimidine-4,5,6-triamine sulfate hydrate, pyrimidine-4,5,6-triamine sulfuric acid hydrate, pyrimidine-4,5,6-triamine; sulfuric acid; hydrate, zlchem 873, acmc-209nvb, c4h7n5.h2o.h2so4, 4,5,6-triaminopyrimidine sulfate, hydrate
PubChem CID 201714
IUPAC Name pyrimidine-4,5,6-triamine;sulfuric acid;hydrate
SMILES OS(O)(=O)=O.NC1=NC=NC(N)=C1N

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 223.22 (Anhydrous)
Percent Purity ≥98%
Assay (dry wt.), water <8%
Chemical Name or Material 4,5,6-Triaminopyrimidine sulfate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.