missing translation for 'onlineSavingsMsg'
Learn More

4-(4'-Methoxy-4-biphenylylsulfonylamino)benzoic acid, 96%, Thermo Scientific™

Product Code. 15498205
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15498205 1 g 1g
15408215 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15498205

Brand: Thermo Scientific Alfa Aesar H32828.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885269-42-1
Molecular Formula C20H17NO5S
Molecular Weight (g/mol) 383.418
MDL Number MFCD06409458
InChI Key ZFNQOUZSNZLJLR-UHFFFAOYSA-N
Synonym 4-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid, 4-4'-methoxy-4-biphenylylsulfonylamino benzoic acid
PubChem CID 44669168
IUPAC Name 4-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]benzoic acid
SMILES COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O

Specifications

Melting Point >250°C
Quantity 1 g
Formula Weight 383.43
Percent Purity 96%
Chemical Name or Material 4-(4'-Methoxy-4-biphenylylsulfonylamino)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.