missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-n-nonyl-2,2'-bipyridine, 97%

Product Code. 15474097
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15474097

Brand: Thermo Scientific Alfa Aesar H31476.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142646-58-0 | C28H44N2 | 408.674 g/mol

Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142646-58-0
Molecular Formula C28H44N2
Molecular Weight (g/mol) 408.674
MDL Number MFCD00800915
InChI Key VHJFWJXYEWHCGD-UHFFFAOYSA-N
Synonym 4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl
PubChem CID 4285174
IUPAC Name 4-nonyl-2-(4-nonylpyridin-2-yl)pyridine
SMILES CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC

Specifications

Melting Point 61°C to 63°C
Quantity 5 g
Formula Weight 408.67
Percent Purity 97%
Chemical Name or Material 4,4'-Di-n-nonyl-2,2'-bipyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.