Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Vo29
Product Code. 16354557 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 16354557
Brand: Memmert™ VO29V6
Only null left
Add to basket
Add to basket
Memmert™ Temperature Output for external temperature monitoring.
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
For Use With (Equipment) Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Type Current Loop Interface
Amperage 4 to 20 mA