Learn More
Memmert™ 4-20 mA Current Loop Interface for UN160plus, Actual Value
Product Code. 17529945 Shop All Memmert Products
Change view
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Amperage unitSize
17529945 4 to 20 mA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17529945 Supplier Memmert™ Supplier No. UN160plusV3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) UN160plus Oven
Type Current Loop Interface
Amperage 4 to 20 mA