Learn More
4-(2-Methylphenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™
Product Code. 15478525
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15478525
Brand: Thermo Scientific Alfa Aesar H33556.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610277-83-3
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.74
MDL Number MFCD01631900
InChI Key IJPKGMOVNZHZKZ-UHFFFAOYSA-N
Synonym 4-2-methylphenoxy benzenesulfonyl chloride, 4-o-tolyloxyphenylsulfonyl chloride, 4-o-tolyloxy-benzenesulfonyl chloride, 4-o-tolyloxy benzenesulfonyl chloride, 4-o-tolyl-oxy-benzenesulfonyl chloride, 4-o-tolyloxy benzene-1-sulfonyl chloride, 4-2-methylphenoxy benzenesulphonyl chloride, 4-2-methylphenoxy benzene-1-sulfonyl chloride, 4-2-methylphenoxy phenylsulfonyl chloride, 4-2-methylphenoxy benzenesulfonylchloride
PubChem CID 2794712
IUPAC Name 4-(2-methylphenoxy)benzenesulfonyl chloride
SMILES CC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O
Specifications
Melting Point 81°C to 85°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 282.74
Percent Purity 96%
Chemical Name or Material 4-(2-Methylphenoxy)benzenesulfonyl chloride