Learn More
4-[2-(Dimethylamino)ethoxy]benzaldehyde, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250mg
1g
5g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Synonym | 4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde |
Quantity | 250mg |
Formula Weight | 193.24 |
Percent Purity | 95% |
Chemical Name or Material | 4-[2-(Dimethylamino)ethoxy]benzaldehyde |