Learn More
4-(2-Aminoethyl)benzenesulfonamide, 99%, Thermo Scientific™
Product Code. 11465606
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11465606
Brand: Thermo Scientific Alfa Aesar B20745.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35303-76-5
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD00010301
InChI Key FXNSVEQMUYPYJS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonamide, 4-2-aminoethyl benzene sulfonamide, 4-2-aminoethyl benzene-1-sulfonamide, 4-2-amino-ethyl-benzenesulfonamide, benzenesulfonamide, 4-2-aminoethyl, 4-2-aminoethyl benzenesulphonamide, chembl7087, 4-aminoethylbenzenesulfonamide, 2-4-sulfamoylphenyl ethylamine
PubChem CID 169682
IUPAC Name 4-(2-aminoethyl)benzenesulfonamide
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)N
Specifications
Melting Point 150°C to 152°C
Quantity 25 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 200.26
Percent Purity 99%
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonamide