missing translation for 'onlineSavingsMsg'
Learn More

4,17-Dioxabicyclo[12.3.2]nonadecane-18-O-tert-butyldimethylsilyl N-Desmethyl Spiramycin I 2A-Acetate, TRC

Product Code. 30385464
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385464 50 mg 50mg
30378394 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385464 Supplier Toronto Research Chemicals Supplier No. TRCD48526050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,17-Dioxabicyclo[12.3.2]nonadecane-18-O-tert-butyldimethylsilyl N-Desmethyl Spiramycin I 2A-Acetate
Analyte or Component Names 4,17-Dioxabicyclo[12.3.2]nonadecane-18-O-tert-butyldimethylsilyl N-Desmethyl Spiramycin I 2A-Acetate
Shipping Condition Room Temperature
Molecular Formula C50 H88 N2 O15 Si
Synonym [9(2R,5S,6R)]-18-Deoxo-3-deoxy-9-O-[5-(methylamino)tetrahydro-6-methyl-2H-pyran-2-yl]-18-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-3,18-epoxy-leucomycin V Acetate
InChI Formula InChI=1S/C50H88N2O15Si/c1-28-24-34-25-40(67-68(15,16)49(7,8)9)64-37(26-38(54)58-29(2)20-18-17-19-21-36(28)63-39-23-22-35(51-11)30(3)59-39)45(57-14)44(34)66-48-46(62-33(6)53)42(52(12)13)43(31(4)61-48)65-41-27-50(10,56)47(55)32(5)60-41/h17-19,21,28-32,34-37,39-48,51,55-56H,20,22-27H2,1-16H3/b18-17+,21-19+/t28-,29-,30-,31-,32+,34-,35+,36+,37-,39+,40?,41+,42+,43-,44+,45+,46-,47+,48+,50-/m1/s1
Solution Type Neat
SMILES CN[C@H]1CC[C@H](O[C@H]2\C=C\C=C\C[C@@H](C)OC(=O)C[C@H]3OC(C[C@@H](C[C@H]2C)[C@H](O[C@@H]4O[C@H](C)[C@@H](O[C@H]5C[C@@](C)(O)[C@@H](O)[C@H](C)O5)[C@@H]([C@H]4OC(=O)C)N(C)C)[C@H]3OC)O[Si](C)(C)C(C)(C)C)O[C@@H]1C
IUPAC Name [(2S,3R,4S,5S,6R)-2-[[(1R,5R,7E,9E,11R,12R,14R,18S,19S)-16-[tert-butyl(dimethyl)silyl]oxy-18-methoxy-5,12-dimethyl-11-[(2R,5S,6R)-6-methyl-5-(methylamino)oxan-2-yl]oxy-3-oxo-4,17-dioxabicyclo[12.3.2]nonadeca-7,9-dien-19-yl]oxy]-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-6-methyloxan-3-yl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 985.324
Quantity 50 mg
Formula Weight 984.595
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.