Learn More
Alfa Aesar™ 4-(1,4-Cyclohexadien-1-yl)-1-butanol, 97%, stab. with 0.1% BHT
Product Code. 15469065
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15469065
Brand: Alfa Aesar™ H36058.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 956150-13-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD22682825
InChI Key QUBPGTNRIJGDFP-UHFFFAOYSA-N
Synonym 4-cyclohexa-1,4-dien-1-yl butan-1-ol, 4-1,4-cyclohexadien-1-yl-1-butanol
PubChem CID 57103928
IUPAC Name 4-cyclohexa-1,4-dien-1-ylbutan-1-ol
SMILES C1C=CCC(=C1)CCCCO
Specifications
Density 0.957
Boiling Point 247°C to 248°C
Flash Point 103°C (217°F)
Refractive Index 1.503
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 152.23
Percent Purity 97%
Chemical Name or Material 4-(1,4-Cyclohexadien-1-yl)-1-butanol, stabilized with 0.1% BHT