Learn More
3M™ Versaflo™ TR-300 / Speedglas™ TR-300 SG PAPR Accessory, Airflow Indicator
Description
The TR-971 air flow indicator may be used to confirm air flow levels of the TR-600 series PAPR. The TR-971 is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system. See the User Instructions for details regarding how to use the TR-971.
- Vortemp Air-Heating valve
- ADF not included
- 3M Catalog# TR-970
Specifications
Specifications
| Disposable | Reusable |
| Product Type | Airflow Indicator |
| For Use With (Application) | Ideal to Confirm Air Flow Levels and to Enable Testing at Various Elevations and Temperatures Within The Operational Range of The Powered Air Purifying Respirator System |
| For Use With (Equipment) | Versaflo TR-300, Speedglas TR-300 SG PAPR System |
| Model | TR-970 |