Learn More
3M KCl Electrolyte Solution, Hamilton™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
500mL
Each
Description
- Helps with buffer values changing over time
Specifications
Specifications
| Chemical Name or Material | EasyFerm Plus VP 425 |
| pH | 0 to 14 |
| Quantity | 500 mL |