Learn More
3M™ Exhalation Valve Replacement Part, 6500 Series Half Mask Respirator
Click to view available options
:
Exhalation valve
Description
- Enhanced comfort by reducing heat and moisture
- Easier breathing with unique valve design
- Replacement part for the 3M™ 6500 Series
- 10 each per case
- For occupational use only
Specifications
Specifications
| Connection Type | Bayonet |
| Color | White |
| Product Type | Exhalation Valve |
| For Use With (Equipment) | Reusable Half Masks 6500 Series |
| Model | 6583 |