missing translation for 'onlineSavingsMsg'
Learn More

3CAI, MedChemExpress

Product Code. 16497108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497108 10 mg 10mg
16483147 50 mg 50mg
16409657 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497108 Supplier Medchem Express Supplier No. HY16666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3CAI is a potent and specific AKT1 and AKT2 inhibitor.

3CAI is a potent and specific AKT1 and AKT2 inhibitor.

Specifications

Chemical Name or Material 3CAI
Physical Form Solid
CAS 28755-03-5
Color Pink
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H8ClNO
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 250 mg/mL (1291.12 mM; Need ultrasonic)
SMILES O=C(CCl)C1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 193.63
Formula Weight 193.63
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.