Learn More
Brannan™ ECO-therm Lab Thermometer
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-35°C to +50°C
Immersion Type:
Partial Immersion
Total Immersion
Unit Size:
Pack of 10
Specifications
Specifications
| Immersion Type | Partial Immersion |
| Length (Metric) | 305 mm |
| Material | Glass |
| Thermometer Fill | Green Spirit |
| Product Type | Laboratory Immersion Thermometer |
| Max. Temperature (Metric) | 50°C |
| Min. Temperature (Metric) | -35°C |
| Temperature Range (Metric) | -35°C to +50°C |
| Accuracy | ±1.5°C |
| Subdivision | 1°C |