Learn More
Watson-Marlow Pumps 318MC Series Pumphead
Product Code. 12821921
Click to view available options
Model:
318MC
318MCX
Unit Size:
Each
Product Code. 12821921
Brand: Watson-Marlow Pumps 033.6853.000
Only null left
Add to basket
Add to basket
318MC Pumphead features high accuracy microcassettes for manifold tubing for flow rates up to 53 mL/min.
  • Designed for multi-channel low-flow applications.
  • For simple tube change without disturbing other channels, cassettes are pre-loaded with tubing, which is available in twenty sizes and three different materials.
  • Extension pumpheads snap fit in seconds, and two MCX (five channel) or three MCX3 (three channel extension pumpheads can be added), depending on the power of the drive.
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 36 mL/min.
Model 318MC
Type Multi-channel Pump Head
Rotor Material Electroless Nickel Plated Hardened Steel, Glass Filled Nylon
Case Material Glass Filled Polypropylene, IXEF (Polyarylamide), Stainless Steel