Learn More
Nickel Electro™ Stainless Steel Laboratory Funnel
Product Code. p-8013865 Shop All Nickel-Electro Products
Click to view available options
Diameter (Metric)Top:
103 mm
165 mm
210 mm
Length (Metric) Stem:
55 mm
75 mm
90 mm
Height (Metric):
115 mm
158 mm
205 mm
Unit Size:
Each
Product Code. 12963621
Brand: Nickel Electro™ FUNSS100H
Use with or without filter membranes for gravity filtration as well as simple funnel procedures. Nickel Electro™ Stainless Steel Laboratory Funnel is vented to allow complete, uninhibited flow of liquids or solids.
  • Durable stainless steel funnel, break-resistant, stain proof and easy to clean.
  • May be used with or without filter membranes for gravity filtration as well as simple funnel procedures.
  • Flute is vented to allow complete, uninhibited flow.
  • Can be used with liquids or solids.
  • The handle can also be used as a convenient hanging point when not in use.
  • Smooth finish on internal surfaces making funnel easy to clean or sterilise.
Specifications
Diameter (Metric)Top 103 mm
Material Stainless Steel
Height (Metric) 115 mm
Includes Handle
Length (Metric) Stem 55 mm