missing translation for 'onlineSavingsMsg'
Learn More

Dopamine-D2 Hydrochloride, TRC

Product Code. 30386059
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386059 10 mg
30379105 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386059 Supplier Toronto Research Chemicals Supplier No. TRCD53378310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27160-01-6
Molecular Formula C8 D2 H9 N O2 . Cl H
Molecular Weight (g/mol) 191.652
Synonym 1,2-Benzenediol, 4-(2-aminoethyl-1,1-d2)-, hydrochloride (9CI), Pyrocatechol, 4-(2-aminoethyl-1,1-d2)-, hydrochloride (8CI), Dopamine-d2 hydrochloride, Dopamine-D2 (2-aminoethyl-1,1-D2) hydrochloride, 4-(2-Aminoethyl-1,1-d2)-1,2-benzenediol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethyl-2,2-d2-amine HCl
IUPAC Name 4-(2-amino-1,1-dideuterioethyl)benzene-1,2-diol;hydrochloride
SMILES Cl.[2H]C([2H])(CN)c1ccc(O)c(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i3D2;
Formula Weight 191.068
Chemical Name or Material Dopamine-D2 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.