missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Donepezil (Mixture of Diastereomers), TRC

Product Code. 30381043
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381043 50 mg -
30386434 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381043 Supplier Toronto Research Chemicals Supplier No. TRCD44909550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120012-04-6
Molecular Formula C24 H31 N O3
Molecular Weight (g/mol) 381.51
Synonym 1H-Inden-1-ol, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, 2,3-Dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-1H-inden-1-ol
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydro-1H-inden-1-ol
SMILES COc1cc2CC(CC3CCN(Cc4ccccc4)CC3)C(O)c2cc1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H31NO3/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18/h3-7,14-15,17,20,24,26H,8-13,16H2,1-2H3
Formula Weight 381.2304
Chemical Name or Material Dihydro Donepezil (mixture of diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.