missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Diaminouracil Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 5,6-Diaminouracil Monosulfate |
| CAS | 886762-39-6 |
| Molecular Formula | C6 H4 Cl2 F N |
| Quantity | 25 g |
| Synonym | 2,4(1H,3H)-Pyrimidinedione, 5,6-diamino-, sulfate (1:1), 5,6-Diamino-2,4-dihydroxypyrimidine sulfate, 5,6-Diaminouracil sulfate, NSC 264289, NSC 3102, NSC 42280, 5,6-Diaminouracil monosulfate |
| InChI Formula | InChI=1S/C6H4Cl2FN/c7-3-1-2-4(10)6(9)5(3)8/h1-2H,10H2 |
| SMILES | Nc1ccc(Cl)c(Cl)c1F |
| IUPAC Name | 3,4-dichloro-2-fluoroaniline |
| Molecular Weight (g/mol) | 180.007 |
| Formula Weight | 178.97 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?