missing translation for 'onlineSavingsMsg'
Learn More

7-Dehydroxy Buprenorphine (Buprenorphine Impurity F), TRC

Product Code. 30348865
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348865 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348865 Supplier Toronto Research Chemicals Supplier No. TRCD23063510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Dehydroxy Buprenorphine
CAS 97203-04-8
Molecular Formula C29 H39 N O3
Quantity 10 mg
Synonym (5α,7α)-17-(Cyclopropylmethyl)-7-(2,2-dimethyl-1-methylenepropyl)-4,5-epoxy-18,19-dihydro-6-methoxy-6,14-ethenomorphinan-3-ol, 17-(Cyclopropylmethyl)-4,5α-epoxy-6-methoxy-7α-[1-(1,1-dimethylethyl)ethenyl]-6α,14-ethano-14α-morphinan-3-ol, Buprenorphine Imp. F (EP)
InChI Formula InChI=1S/C29H39NO3/c1-17(26(2,3)4)20-15-27-10-11-29(20,32-5)25-28(27)12-13-30(16-18-6-7-18)22(27)14-19-8-9-21(31)24(33-25)23(19)28/h8-9,18,20,22,25,31H,1,6-7,10-16H2,2-5H3/t20-,22-,25-,27-,28+,29?/m1/s1
SMILES CO[C@]12CC[C@@]3(C[C@@H]1C(=C)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4CC7CC7)c56
IUPAC Name (1S,2S,6R,14R,15R,16R)-5-(cyclopropylmethyl)-16-(3,3-dimethylbut-1-en-2-yl)-15-methoxy-13-oxa-5-azahexacyclo[13.2.2.12,8.01,6.02,14.012,20]icosa-8(20),9,11-trien-11-ol
Molecular Weight (g/mol) 449.62
Formula Weight 449.293
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.