missing translation for 'onlineSavingsMsg'
Learn More

12-Amino-1-dodecanoic Acid Methyl Ester Hydrochloride Salt, TRC

Product Code. 30312458
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312458 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312458 Supplier Toronto Research Chemicals Supplier No. TRCA6088071G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-Amino-1-dodecanoic Acid, Methyl Ester, Hydrochloride Salt
CAS 4271-86-7
Molecular Formula C13 H27 N O2 . Cl H
Quantity 1 g
InChI Formula InChI=1S/C13H27NO2.ClH/c1-16-13(15)11-9-7-5-3-2-4-6-8-10-12-14;/h2-12,14H2,1H3;1H
SMILES Cl.COC(=O)CCCCCCCCCCCN
IUPAC Name methyl 12-aminododecanoate;hydrochloride
Molecular Weight (g/mol) 265.82
Formula Weight 265.1809
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.