Learn More
Skytec™ 3020 Disposable FFP2 Pre-Formed Cup Shape Respirator
Click to view available options
:
FFP2
Description
- Lightweight, cup shaped respirator
- Continuous loop metal-free headstrap
- Preformed nosebridge removes the need for regular adjustment
- Conforms to EN 149: 2001 + A1: 2009 FFP2 NR
Specifications
Specifications
| Assigned Protection Factor (APF) | 10 |
| Certifications/Compliance | EN 149:2000 + A1:2009 FFP2 NR |
| Color | White |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | FFP2 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | No |
| Show More |