missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ 30 mL Diamond Weighing Boat

Product Code. p-7182159
Click to view available options
Color:
Black
White
Unit Size:
Pack of 5000
This item is not returnable. View return policy

Product Code. 16405019

Brand: Wheaton™ DS30B/AS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Weighing boat with a smooth surface and rounded corners that allow accurate weighing and controlled pouring of liquid, powdered, or granular samples.

  • Anti-static
  • Prevents any sample retention
  • Diamond-shaped bottom
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bottom Shape Diamond
Capacity (Metric) 30 mL
Color Black
Disposable Single-use
Material High-impact Polystyrene
Sterility Non-sterile
For Use With (Equipment) Weighing Scales
No. per Case 5000
No. per Pack 500
Dimensions (L x W) 85 x 65 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.