missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%

Product Code. 11408826
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11408826

Brand: Thermo Scientific Alfa Aesar A16983.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 779-89-5 | C10H7F3O2 | 216.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-89-5
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.159
MDL Number MFCD00004393
InChI Key KSBWHDDGWSYETA-SNAWJCMRSA-N
Synonym 3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid, m-trifluoromethyl cinnamic acid, e-3-trifluoromethyl cinnamic acid, unii-p9lj9on37w, 2e-3-3-trifluoromethyl phenyl prop-2-enoic acid, e-m-trifluoromethyl cinnamic acid, 3-e-trifluoromethyl cinnamic acid
PubChem CID 719451
ChEBI CHEBI:60704
IUPAC Name (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)O

Specifications

Melting Point 132°C to 136°C
Odor Odorless
Quantity 25 g
Beilstein 1963372
Formula Weight 216.16
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)cinnamic acid, predominantly trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.