Learn More
Fisherbrand™ 3-Ply Sterile Facemasks
Product Code. p-9818002 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
17 cm
23 cm
Unit Size:
Pack of 200
Product Code. 17307299
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3-Ply Sterile Facemasks constructed of polyethylene apertured film and PTFE (polytetrafluoroethylene).
  • Constructed from a combination of pleated clear, polyethylene apertured film and PTFE (polytetrafluoroethylene), with soft tie strings
  • Manufactured in systems complying ISO 13485:2016 requirements
  • Certified Sterile
Specifications
Product Type Cleanroom Face Mask
Color White
Sterility Sterile
Style Pleated with Ties
Certifications/Compliance The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013.
Disposable Single-use
For Use With (Application) Industrial Use Only
Length (Metric) 23 cm