missing translation for 'onlineSavingsMsg'
Learn More

3-Oxohexanedioic Acid Diethyl Ester, TRC

Product Code. 30411584
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411584 1 g -
30418756 500 mg 500mg
30417552 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411584 Supplier Toronto Research Chemicals Supplier No. TRCO8568301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7149-59-9
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
Synonym 3-Oxo-hexanedioic Acid 1,6-Diethyl ester, 3-Oxoadipic acid diethyl ester, 3-Oxohexanedioic acid diethyl ester, Diethyl 3-oxohexanedioate, Diethyl 4-oxohexanedioate, Diethyl β-oxoadipate, NSC 72263
IUPAC Name diethyl 3-oxohexanedioate
SMILES CCOC(=O)C(C)(C)Cc1ccc(S)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H16O5/c1-3-14-9(12)6-5-8(11)7-10(13)15-4-2/h3-7H2,1-2H3
Formula Weight 216.0998
Chemical Name or Material 3-Oxohexanedioic Acid Diethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.