missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Oxobutanenitrile, 96%
Click to view available options
Quantity:
250 mg
1 g
Tamaño de la unidad:
1g
250mg
Description
It is used as pharmaceutical raw material and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Color | Colorless to Yellow |
| Boiling Point | 92°C to 94°C (10 mmHg) |
| Linear Formula | CH3COCH2CN |
| Refractive Index | 1.4285 |
| Quantity | 250 mg |
| UN Number | UN3276 |
| Beilstein | 1633584 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 83.09 |
| Percent Purity | 96% |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction