missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-O-(2-Aminoethyl)-25-hydroxyvitamin D3, MedChemExpress
Description
3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative.
Specifications
Specifications
| Chemical Name or Material | 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 |
| CAS | 163018-26-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H51NO2 |
| Quantity | 10 mg |
| Synonym | 25-Hydroxy Vitamin D3 3,3’-Aminopropyl Ether |
| Solubility Information | DMSO : ≥ 100 mg/mL (218.47 mM) |
| SMILES | C=C1CC[C@H](OCCCN)C/C1=C/C=C2[C@@](CC[C@]3([H])[C@@H](CCCC(C)(C)O)C)([H])[C@]3(C)CCC/2 |
| Molecular Weight (g/mol) | 457.73 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?