missing translation for 'onlineSavingsMsg'
Learn More

3-O-(2-Aminoethyl)-25-hydroxyvitamin D3, MedChemExpress

Product Code. 16471489
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471489 1 mg 1mg
16496398 10 mg 10mg
16478987 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471489 Supplier Medchem Express Supplier No. HY15254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative.

3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative.

Specifications

Chemical Name or Material 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3
CAS 163018-26-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H51NO2
Quantity 1 mg
Synonym 25-Hydroxy Vitamin D3 3,3’-Aminopropyl Ether
Solubility Information DMSO : ≥ 100 mg/mL (218.47 mM)
SMILES C=C1CC[C@H](OCCCN)C/C1=C/C=C2[C@@](CC[C@]3([H])[C@@H](CCCC(C)(C)O)C)([H])[C@]3(C)CCC/2
Molecular Weight (g/mol) 457.73
Formula Weight 457.73
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.